missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALPK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555763
This item is not returnable.
View return policy
Description
Immunogen sequence: EKPTTLTANN ECFQATRETE DTSTVTIATE VHPAKYLAVS IPEDKHAGGT EERFPRASHE KVSQFPSQVQ VDHILSGATI KSTKELLCRA PSVPGVP Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 37%.
Protein kinase that recognizes phosphorylation sites in which the surrounding peptides have an alpha-helical conformation (PubMed:10021370). Regulates cardiac development and cardiomyocyte differentiation by negatively regulating Wnt/beta-catenin signaling (PubMed:29888752). [UniProt].
Specifications
| ALPK2 | |
| Polyclonal | |
| Unconjugated | |
| ALPK2 | |
| alpha kinase 2; alpha-kinase 2; alpha-protein kinase 2; Alpk2; Gm549; HAK; heart alpha-kinase; heart alpha-protein kinase; RGD1564431; RNF18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 115701 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86TB3 | |
| ALPK2 | |
| Recombinant protein corresponding to Human ALPK2. Recombinant protein control fragment (Product #RP-95081). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction