missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALPK1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | ALPK1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18359831
|
Bio-Techne
NBP3-15997-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18320792
|
Bio-Techne
NBP3-15997-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALPK1 Polyclonal antibody specifically detects ALPK1 in Human samples. It is validated for Western BlotSpecifications
| ALPK1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 80216 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| alpha-kinase 1, alpha-protein kinase 1, Chromosome 4 kinase, EC 2.7.11.-, KIAA15278430410J10Rik, Lak, LAKFLJ22670, lymphocyte alpha-kinase, Lymphocyte alpha-protein kinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 340-500 of human ALPK1 (NP_001095876.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title