missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALPK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19107-1-AP-150UL
This item is not returnable.
View return policy
Description
Unlike most eukaryotic kinases, alpha kinases, such as LAK, recognize phosphorylation sites in which the surrounding peptides have an alpha-helical conformation.Specifications
| ALPK1 | |
| Polyclonal | |
| Unconjugated | |
| ALPK1 | |
| 8430410J10Rik, alpha kinase 1, Alpha protein kinase 1, ALPK1, Chromosome 4 kinase, KIAA1527, LAK | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 310879, 71481, 80216 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96QP1, Q9CXB8 | |
| Alpk1 | |
| ALPK1 Fusion Protein Ag5320 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction