missing translation for 'onlineSavingsMsg'
Learn More

Alphazurine A, MP Biomedicals™

Product Code. 11484761 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484761 25 g 25g
11494761 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11484761 Supplier MP Biomedicals Supplier No. 0215473025
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Soluble in water (1mg/mL); Store at Room Temperature (15° to 30°C)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3486-30-4
Molecular Formula C37H35N2NaO6S2
Molecular Weight (g/mol) 690.805
MDL Number MFCD00044440
InChI Key FTUYQIPAPWPHNC-UHFFFAOYSA-M
Synonym acid blue 7, alphazurine a, patent blue a, fenazo bluexg, erio glaucine x, hidacid blue a, carmine blue af, hidacid blue af, kiton blue a, patent blue af
PubChem CID 62498
IUPAC Name sodium;4-[[4-[benzyl(ethyl)amino]phenyl]-[4-[benzyl(ethyl)azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzene-1,3-disulfonate
SMILES CCN(CC1=CC=CC=C1)C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC=CC=C4)C=C3)C5=C(C=C(C=C5)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+]

Specifications

Color Purple
Quantity 25 g
Formula Weight 690.8
Physical Form Powder
Chemical Name or Material Alphazurine A
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.