Learn More
Ansell™ AlphaTec™MICROCHEM™4000 Model 215 Apron with Sleeves
Product Code. 15875342 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 20
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15875342 Large Pack of 20
15855342 Small Pack of 20
15865342 Medium Pack of 20
15885342 2X-Large Pack of 20
15895342 3X-Large Pack of 20
15805352 4X-Large Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15875342 Supplier Ansell™ Supplier No. GR40T0021504
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
MICROCHEM™ 4000 is designed to provide anexceptional barrier against organic and inorganicchemicals and biological agents
  • Velcro fastening to neck
  • Tie fastening at the waist
  • Double cuff with knitted
  • inner cuff
  • Welded and taped seams
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Large
Material Multi-layer Barrier Laminate Fabric
Color Green
Disposable Limited-use
Closure Type Hook and Loop, Waist Ties
Sterility Non-sterile
Cuff Style Double Inner Knit
Latex Free Yes
No. of Pockets None
Resistance Abrasions, acids, anti-static, bloodborne pathogens, chemicals
Certifications/Compliance EN 14605: 2005 +A1:2009, Type PB (3); EN 14605: 2005 +A1:2009, Type PB (4); EN14126: 2003, Types PB(3)-B, PB (4)-B
Style Alphatec™ 4000, Model 215
Seam Type Welded and Taped