missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- AlphaTec™ 58-001 ESD - Unique Nitrile and Carbon formulation consistently dissipates static across the surface and through the glove ensuring consistent passing of EN 1149 and EN 16350
- High performance
- Nitrile compound offers a far superior snag, puncture and abrasion protection compared with rubber or Neoprene gloves
- Reverse lozenge palm finish for improved dry/wet grip
- Foldable cuff gutter to prevent dripping on the forearm
- Antistatic flocking layer for ease of donning/doffing and moisture management
- For Assembly and inspection of components, Oil, fluids and filter change, Plating, coating, sealing, painting, Handling incoming goods, Transferring liquids and solids between vessels and tanks and process equipment, Opening furnaces draining pumps valves or lines and crackers BTX process, Cleaning furnaces, distillation pumps, valves or lines and crackers BTX process
Specifications
Specifications
| ANSI Abrasion Rating | ANSI 4 |
| Certifications/Compliance (Europe) | EN 388:2016 +A1:2018, EN 420:2003 + A1:2009, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-5:2016, EN 1149, EN 16350:2014, EN 16350:2014, EN 388:2016, EN ISO 374-1:2016 |
| Accelerator Free | No |
| Applications | Industrial |
| Coating Material | Nitrile |
| Color | Green |
| Cut Resistance Rating | ANSI A1 |
| Disposable | Reusable |
| Exterior Finish | No Texture |
| Hand | Ambidextrous |
| Show More |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?