Learn More
Ansell AlphaTec™ Airline Passthrough without Connector
Product Code. 18813796
Change view
Click to view available options
Includes:
Without Connector
Product Code. Includes
18813796 Without Connector
1 options
Product Code. 18813796 Supplier Ansell Supplier No. AC01P0002200N00
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Provides external air supply to AlphaTec reusable gas-tight suits, for breathing air as well as suit ventilation.
  • For use with external air supply: Useful during suit decontamination
  • Large thumbwheel: Good grip, easy adjustments
  • Compatible with all SCBA brands: Improved practicality and convenience
TRUSTED_SUSTAINABILITY
Specifications
Flow Rate 100 L/min.
Material Glass Fiber Reinforced PPS, Polyamide, Anti-static Polyester
Includes Without Connector
Product Type Air Supply Connector