missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 87-085 Lightweight Food-Safe Latex Gloves

Product Code. p-7215039 Shop All Ansell Products
Click to view available options
Size:
6.5
7.5
8.5
9.5
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 16845250

missing translation for 'mfr': Ansell 87085065BP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves are ideal for low hazard areas, and feature a lightweight flocked lining for comfort.

  • For use in low hazard areas
  • Beaded cuff for tear resistance and easy donning
  • Resistant, flexible, and comfortable
  • 100% cotton flock lining
  • AQL 0.65
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, REACH Compliant
Color Pink
Hand Specific
Material Latex
Size 6.5
EN388 Standard 2010X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KLT
EN ISO 374 - 5 Yes
Liner Material 100% Cotton
Series 87-085
Length (English) 12 in.
Length (Metric) 300 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.