missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 87-085 Lightweight Food-Safe Latex Gloves

Product Code. 16875250 Shop All Ansell Products
Change view
Click to view available options
Size:
6.5
7.5
8.5
9.5
Unit Size:
12 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16875250 9.5 12 pairs
16845250 6.5 12 pairs
16855250 7.5 12 pairs
16865250 8.5 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16875250 Supplier Ansell Supplier No. 87085095
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Gloves are ideal for low hazard areas, and feature a lightweight flocked lining for comfort.

  • For use in low hazard areas
  • Beaded cuff for tear resistance and easy donning
  • Resistant, flexible, and comfortable
  • 100% cotton flock lining
  • AQL 0.65
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, REACH Compliant
Color Pink
Hand Specific
Material Latex
Size 9.5
EN388 Standard 2010X
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KLT
EN ISO 374 - 5 Yes
Liner Material 100% Cotton
Series 87-085
Length (English) 12 in.
Length (Metric) 300 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.