missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 87-029 Protective Gloves

Product Code. 30286223 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
120 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30286223 8 12 pairs
30286206 7 12 pairs
30286208 9 12 pairs
30286203 10 12 pairs
18835584 11 120 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30286223 Supplier Ansell Supplier No. 87198080
In Stock
Add to basket
Add to basket

Anatomically designed natural rubber gloves, with curved fingers and a seamless nylon liner.

  • Natural latex provides a high degree of tensile strength
  • Cotton flock lining for best comfort
  • Good chemical and mechanical protection
  • Special wax treated for water repelling
  • Excellent dexterity and tactility
  • Compliant to FDA and EU food safety requirements (including 21 CFR 177.2600)
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Color Lemon Yellow
Finger Thickness (English) 15.7 mil
Finger Thickness (Metric) 0.4 mm
Latex Free Yes
Material Rubber Latex
Palm Thickness (English) 15.7 mil
Palm Thickness (Metric) 0.4 mm
Product Type Chemical Protection Gloves
Size 8
Thickness (English) 15.7 mil
Thickness (Metric) 0.4 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.