missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 58-535B Series Nitrile Immersion Gloves
Click to view available options
Size:
7
8
9
10
11
Description
Provides dexterity with resistance to cuts, punctures, and abrasions as well as petrochemicals, oils, greases, pesticides, and other agricultural chemicals
- Chemical resistant
- Material fully coated nitrile gloves
- Thickness: 11 mil
- Embossed grip surface for superior wet and oil grip
- Red with black coating
- Comfortable design reduces fatigue
Specifications
Certifications/Compliance | FDA 21 CFR 177-2600 |
Color | Green, Black |
Latex Free | Yes |
Material | Nitrile |
Size | 9 |
Thickness (English) | 13 mil |
Thickness (Metric) | 0.33 mm |
For Use With (Application) | Chemical plants and transportation, chemical processing, oil refining and petrochemicals, cleaning and de-greasing, exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, janitorial and maintenance work |
Glove Cuff Style | Gauntlet |
Liner Material | Knit Lined Acrylic |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'