missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 58-530W Series Nitrile Immersion Gloves

Product Code. 12649741 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
6 pairs
Product Code. Size unitSize
12649741 10 6 pairs
12659741 11 6 pairs
12669741 7 6 pairs
12679741 8 6 pairs
12689741 9 6 pairs
5 options

Product Code. 12649741

Brand: Ansell™ 58535W100

Only null left
Add to basket
Add to basket

Recommended for outdoor applications. Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture to prevent possible chemical leakage onto the skin.

  • Nitrile polymer coating reduces chemical leakage onto skin
  • For indoor applications
  • Nylon liner
  • Gauntlet cuff
  • Sea green shell, anthracite gray dip color
TRUSTED_SUSTAINABILITY

Specifications

Back Material Nitrile
Cuff Material Nitrile
Applications Industrial
Coating Material Nitrile
Color Green, Gray
Latex Free Yes
Material Nitrile
Product Type Immersion Gloves
Size 10
Glove Cuff Style Gauntlet
Liner Material Nylon
Lining White Nylon
Quantity 6 Pairs
Resistant To Chemical resistance
Series 58-535W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.