missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 58-201 Green Nitrile Gloves

Product Code. 17923072 Shop All Ansell Products
Click to view available options
Size:
8
9
10
Unit Size:
6 pairs
This item is not returnable. View return policy

Product Code. 17923072

Brand: Ansell 58201100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Utilizing a unique formulation, AlphaTec 58-201 is a truly versatile protective solution that can be applied to a wide range of applications and environments.

  • AnsellGrip Technology for enhanced grip in Wet and Oily conditions.
  • Designed to protect at temperatures from -40 to +250°C/482°F.
  • Acrylic liner keeps the hands warm in cold conditions and protected in contact heat applications.
  • Nitrile chemical barrier providing protection against a wide range of chemicals.
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 5
Certifications/Compliance ANSI ABR 5, ANSI Cut A2, CE 0493, EN 388:2016 +A1:2018, EN 374:2003, EN 388:2003, EN407:2020 , EN 511:2006, Food Contact
Applications Industrial
Color Green
Latex Free Yes
Material Acrylic, Nitrile
Product Type Chemical-resistant Gloves
Size 10
Glove Cuff Style Gauntlet
Grip Design Ansell Grip Technology
Liner Material Acrylic
Length (Metric) 330 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.