missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 58-005 Green Nitrile Chemical Protection Gloves, Nylon Lining

Product Code. 17895631 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
12
Unit Size:
6 pairs
This item is not returnable. View return policy

Product Code. 17895631

Brand: Ansell 58005110

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Optimized fit that integrates the liner into nitrile shell for better dexterity and tactility versus double gloving. Cuff can be folded to act as a gutter to prevent chemicals running down the arm.

  • Unique Nitrile/Neoprene blend provides enhanced chemical permeation and degradation resistance
  • AlphaTec 58-005 has Type A chemical protection under new ENISO 374:2016
  • Ansell Grip Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 388:2016, EN 388:2016 +A1:2018, EN 407:2004, EN407:2020 , EN 420:2003 + A1:2009, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, EN ISO 374-5:2016
Applications Industrial
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 11
EN388 Standard 4122A
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMNOPT
Glove Cuff Style Extended
Liner Material Nylon
Lining Lined
Model 58-005
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.