missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 58-005 Green Nitrile Chemical Protection Gloves, Nylon Lining

Product Code. 17865631 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
12
Unit Size:
6 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17865631 8 6 pairs
17855631 7 6 pairs
17875631 9 6 pairs
17885631 10 6 pairs
17895631 11 6 pairs
17805641 12 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17865631 Supplier Ansell Supplier No. 58005080
Only null left
Add to basket
Edge
Add to basket

Optimized fit that integrates the liner into nitrile shell for better dexterity and tactility versus double gloving. Cuff can be folded to act as a gutter to prevent chemicals running down the arm.

  • Unique Nitrile/Neoprene blend provides enhanced chemical permeation and degradation resistance
  • AlphaTec 58-005 has Type A chemical protection under new ENISO 374:2016
  • Ansell Grip Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 388:2016, EN 388:2016 +A1:2018, EN 407:2004, EN407:2020 , EN 420:2003 + A1:2009, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN ISO 374-1:2016, EN ISO 374-5:2016
Applications Industrial
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 8
EN388 Standard 4122A
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES AJKLMNOPT
Glove Cuff Style Extended
Liner Material Nylon
Lining Lined
Model 58-005
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.