missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 58-270 Series Nitrile Immersion Gloves

Product Code. p-8004829 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
11791683 10 12 pairs
12640952 6 12 pairs
11701693 11 12 pairs
11761683 7 12 pairs
11771683 8 12 pairs
11781683 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11791683 Supplier Ansell™ Supplier No. 58270100
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

Designed with excellent grip and dexterity for light-to-medium chemical applications.

  • Thin, double-wall nitrile shell with 15-gauge seamless nylon liner
  • Lightweight and comfortable
  • Retains minimal heat, reducing perspiration
  • Liquid-proof
  • Safety cuff shape prevents snagging and itching
TRUSTED_SUSTAINABILITY

Specifications

Back Material Nitrile
Cuff Material Nitrile
Applications Industrial
Color Gray
Latex Free Yes
Material Nitrile
Product Type Immersion Gloves
Size 10
Glove Cuff Color Green
Glove Cuff Style Gauntlet
Liner Material Nylon
Quantity 12 Pairs
Resistant To Chemical resistance
Series 58-270
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.