missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Thin, double-wall nitrile shell with 15-gauge seamless nylon liner
- Lightweight and comfortable
- Retains minimal heat, reducing perspiration
- Liquid-proof
- Safety cuff shape prevents snagging and itching
Specifications
Specifications
| Back Material | Nitrile |
| Cuff Material | Nitrile |
| Applications | Industrial |
| Color | Gray |
| Latex Free | Yes |
| Material | Nitrile |
| Product Type | Immersion Gloves |
| Size | 11 |
| Glove Cuff Color | Green |
| Glove Cuff Style | Gauntlet |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?