missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Unique rubber polymer and plastic laminate technology delivering maximum chemical protection against an extremely broad set of chemical types.
- Unique manufacturing process allows for the glove to be form fitting.
- Raised diamond palm pattern for enhanced grip.
- Unique donning and doffing layer to allow ease of use.
- Anti-static according to EN 1149-3.
- Natural rubber and silicone free.
Specifications
Specifications
| Certifications/Compliance | CE 0493, EN ISO 374-5:2016, EN 388:2016 +A1:2018, EN 421:2010, EN ISO 374-1:2016 |
| Chlorinated | No |
| Description | AlphaTec 53-003 the revolutionary glove that redefines chemical protection combined with integral support for mechanical and heat protection. Our innovative design combines an advanced rubber polymer coating with cutting-edge plastic laminate technology, setting a new benchmark in chemical permeation performance whilst delivering comfort and tactility. |
| Applications | Industrial |
| Color | Black |
| Latex Free | Yes |
| Material | Neoprene |
| Product Type | Neoprene Gloves |
| Size | 9 |
| Thickness (English) | 17 mil |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?