missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 53-001 Nitrile Chemical Resistant Gloves

Product Code. p-8854004 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
6 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16583542 7 6 pairs
16593542 8 6 pairs
16503552 9 6 pairs
16513552 10 6 pairs
16523552 11 6 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16583542 Supplier Ansell Supplier No. 53001070
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Black gloves feature an innovative multi-layer polymer design to offer multi-barrier chemical protection.

  • Innovative multi-layer polymer design - nitrile/neoprene/nitrile layers
  • Provides chemical protection against a wide range of chemicals from acids and bases to hydrocarbons and organic solvent
  • Tested according to EN ISO 374:2016, permeation time > 30 minutes against 13 chemicals out of 18 listed
  • Features the latest MICROCHEM™ chemical barrier technology providing superior protection for use in hazardous environments
  • ANSELL GRIP™ Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
  • Inner soft nylon liner for comfort and increased mechanical protection
  • Oeko-Tex™ Standard 100 Approved
Note:  Size equivalents are as follows: 7 (S), 8 (M), 9 (L), 10 (XL), 11 (2XL).

TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 1149, EN407
Chlorinated No
Applications Industrial
Color Gray, Black
Latex Free Yes
Material Nitrile, Neoprene
Palm Thickness (English) 33 mil
Palm Thickness (Metric) 0.84 mm
Product Type Chemical-resistant Gloves
Size 7
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
EN388 Standard EN 388:3121A
EN 421 No
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ACFGJKLMNOPST
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Gauntlet
Grip Design Ansell Grip Technology
Liner Material Nylon
Lining Lined
Model 53001070-NA
Series AlphaTec™ 53-001
Length (English) 13 in.
Length (Metric) 35 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.