missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Specialized protection:The AlphaTec™ 4000 Ultrasonically Welded and Taped - Model 130 is a robust chemical protective suit, permeation-tested to withstand more than 200 hazardous chemicals, including chemical warfare agents
- Increased strength: This full body protection suit comes with taped and welded seams to grant users added strength and chemical barrier defense
- Heightened visibility: Designed with orange-colored material, it ensures better visibility which can reduce the risk of workplace injuries
- Improved practicality: It also possesses a chinstrap and a thumb loop, which are compatible with a range of respirators and hand protection products
- Optimized fit: Its elasticated two-piece hood, waist, cuffs and ankles simplify the donning and doffing process
- Certified anti-static capabilities: In addition, this anti-static suit complies with EN 1149-5
- CE Category III Type 3/4/5 protection
Specifications
Specifications
| Color | Orange |
| Hood Type | Attached |
| Material | Multi-layer Barrier Laminate Fabric |
| Size | Medium |
| Ankle Style | Elasticated |
| Closure Type | Single zip closure |
| Collar Type | 3-piece hood |
| Cuff Style | Elasticated |
| Resistance | Chemical |
| Seam Type | Ultrasonicaly welded and taped |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?