Learn More
Ansell AlphaTec™ 4000 Trousers
Product Code. 17150695
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17150695 Large Pack of 25
17140695 Medium Pack of 25
17160695 X-Large Pack of 25
17170695 2X-Large Pack of 25
17180695 3X-Large Pack of 25
17130695 Small Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17150695 Supplier Ansell Supplier No. GR40T0030104
Estimated Shipment: 13-05-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
A unique multi-layer barrier fabric renowned for its lightweight, yet robust textile feel and exceptional barrier to organic & inorganic chemicals.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Designed to protect
  • Silicone free
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Gender Unisex
Product Type Chemical-resistant Trousers
Material Multi-layer Barrier Laminate Fabric
Color Green
Seam Type Welded and Taped
Certifications/Compliance (Europe) Partial Body Protection Only, EN 14605:2005 + A1:2009, EN 14605:2005 + A1:2009, EN 14126:2003
Style G40T301
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes