Learn More
Ansell Alphatec™ 4000 Avant2 Airline Ultrasonically Welded and Taped - Model 756-G05
Product Code. p-200080411
missing translation for 'orderingAttributeHoverText'
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
missing translation for 'unitSize'
Pack of 3
Product Code. 17160815
missing translation for 'mfr': Ansell GR40TA0075602G05
Only null left
Add to basket
Add to basket
Alphatec™ 4000 Avant2 Airline Ultrasonically Welded and Taped Chemical Protective Suit - Model 756-G05 with welded and taped seams offer the highest level of defense against liquids and particulates.
  • Advanced defenses: The AlphaTec™ 4000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 protective suit was assessed against more than 200 chemicals, including chemical warfare agents
  • Enhanced protection: This chemical protective suit's welded and taped seams offer the highest level of defense against liquids and particulates
  • Added features: The AlphaTec™ 4000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 756 also features a panoramic visor design, which ensures a good field of vision for the wearer
  • Reduced risk: In addition to a range of sleeve options, this protective body suit also comes with attached socks and an elasticated boot over flap to reduce chemical ingress risk
DESIGN FEATURES:
  • Fully encapsulated air-fed suit that features a permanently built in AlphaTec AVANT STS V1 airline regulator. Suit features include internal air-channel ventilation system double cuffs, finger and thumb loop. Attached boots ends with anti-slip PVC sole version also available and emergency rip cord. Attached glove options:. No glove with finger loops, G02(B) AlphaTec™ 02-100 or G05(D) AlphaTec™ 29-500. Styles with codes ending in A756 include an optional CEJN airline connector, styles with codes ending in 0756 do not.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Color Green
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, blending and charging of raw materials, Transferring liquids and solids, Opening and draining pumps valves or lines, Loading and unloading of proces equipment, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size Small
Ankle Style Attached Socks, Elasticated Boot Over Flap
Closure Type Zipper
Collar Type Hood with a panoramic visor
Disposable Reusable
Gender Unisex
Neck Style Hood with Panoramic Visor
Product Type Chemical Protective Suit
Protection Class CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Seam Type Ultrasonically Welded and Taped
Sterility Non-sterile