Learn More
Ansell AlphaTec 4000 AVANT AIRline
Product Code. p-7228385
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Includes:
Attached Socks and Gloves
Socks and Elasticated Boot Overlap
Socks and Neoprene Gloves
Style:
G40T755
G40T755B
G40T755C
G40T755D
Unit Size:
Pack of 3
Product Code. 17120755
Brand: Ansell GR40T0075502G03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Encapsulated chemical protective suits for use with air-fed/supplied systems.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow).
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress.
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Material Multi-layer Barrier Laminate Fabric
Size Small
Includes Socks and Neoprene Gloves
Seam Type Welded and Taped
Style G40T755C
Weight (Metric) 103 gsm