Learn More
Ansell AlphaTec™ 4000 APOLLO
Product Code. 17120665
Change view
Click to view available options
Size:
Small
3X-Large
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17120665 3X-Large Pack of 4
17110665 Small Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17120665 Supplier Ansell Supplier No. GR40T0012607
Estimated Shipment: 24-06-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Fully encapsulated chemical protective and liquid tight suit with Type 3/4 protection.
  • Designed to protect, rear entry double zip system, attached socks with boot overflap
  • Rear mounted breathing apparatus (BA) pouch - universal fit with most BA
  • Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
  • Attached Ansell Barrier™ gloves, exhalation valves and clear face visor
TRUSTED_SUSTAINABILITY
Specifications
Color Green
For Use With (Application) Contaminated Environments, Chemical Handling, Filling, Blending and Charging of Raw Materials, Transferring Liquids and Solids, Opening and Draining Pumps Valves or Lines, Loading and Unloading of Proces Equipment, Raw Material Sample Collection, Concentration Mixing and Palletisation, Hazmat Response, Assembly and Disassembly of Large Pipes
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size 3X-Large
Product Type Fully encapsulated chemical protective, liquid tight suit, Type 3/4 protection
Seam Type Welded and Taped
Style G40T126B
Weight (Metric) 103 gsm