missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 38-612 (ex ChemTek™ 38-612)

Product Code. p-7146537 Shop All Ansell Products
Click to view available options
Size:
8
9
Unit Size:
1 pair

Product Code. 15886932

Brand: Ansell™ 38612080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Maximum protection from hazardous chemicals

  • The AlphaTec™ 38-612 range provides the best resistance to the most aggressive chemicals – without compromising dexterity or comfort.
  • AlphaTec™ 38-612 offers protection against aliphatic, halogenated and aromatic hydrocarbons as well as concentrated mineral acids.
  • AlphaTec™ 38-612 gloves very flexible and provide excellent dexterity. Designed for comfort – with a natural, curved ergonomic shape and soft feel – they offer easy donning and good grip.
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 1
Chlorinated No
Cut Style Beaded
Applications Industrial
Coating Material Viton/Butyl Polymer
Color Black
Exterior Finish Smooth
Latex Free Yes
Material Butyl Polymer
Product Type Butyl Gloves
Size 8
Thickness (English) 0.0118 in.
Thickness (Metric) 0.3 mm
EN 421 No
EN ISO 374-1 Type A
EN ISO 374 - 5 Bacteria, Fungi only
Glove Cuff Style Beaded
Grip Design Smooth
Lining Unlined
Length (English) 12 in.
Length (Metric) 300 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.