missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 37-501 - Nitrile Gloves

Product Code. 30281116 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
12 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30281116 7 12 pairs
30281111 8 12 pairs
30281112 9 12 pairs
30281113 10 12 pairs
30281115 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30281116 Supplier Ansell Supplier No. 37501070
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Blue medium-duty nitrile gloves, for assured dexterity and chemical resistance

  • Specialized protection: AlphaTec™ 37-501 nitrile gloves ensure personal protection against various solvents and oil-based chemicals, while granting wearers dexterity and moderate abrasion resistance
  • Ventilated comfort: As these flexible work gloves feature open cuffs, they ensure better ventilation, keeping workers' hands cool, dry and comfortable
  • Food handling compliance: AlphaTec™ 37-501 protective gloves also comply with relevant food processing standards, making them ideal food handling gloves
TRUSTED_SUSTAINABILITY

Specifications

Applications Industrial
Color Blue
Finger Thickness (English) 16.5 mil
Finger Thickness (Metric) 0.42 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 16.5 mil
Palm Thickness (Metric) 0.42 mm
Product Type Chemical-resistant Gloves
Size 7
Thickness (English) 16.5 mil
Thickness (Metric) 0.42 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.