Learn More
Ansell™ AlphaTec™ 3000 Overboots Ultrasonically Welded and Taped
Product Code. 31114788 Shop All Ansell Products
Change view
Click to view available options
Color:
Yellow
Product Code. Color
31114788 Yellow
1 options
Product Code. 31114788 Supplier Ansell™ Supplier No. YE30T0040600
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Multi-layer barrier fabric for safety footwear with chemical resistance with all-day comfort, hard-wearing protection
  • The AlphaTec™ 3000 STANDARD Overboots Ultrasonically Welded and Taped - Model 406 grants a protective multi-layer barrier fabric that is extremely effective against inorganic chemicals and biological hazards
  • This protective footwear is highly visible, thanks to its bright yellow color, which helps improve workplace safety in low-light conditions
  • Light and durable by nature, these safety overboots offer a blend of comfort and protection
TRUSTED_SUSTAINABILITY
Specifications
Size One Size
Material Multi-layer Barrier Laminate Fabric
Latex Free Yes
Color Yellow
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Opening
Sole Type Standard
Certifications/Compliance (Europe) CE CATEGORIA III (Tipo PB [3]-B, Tipo PB [4]-B, Tipo PB [6]-B, EN 14126
Resistance Chemical-resistant, Bloodborne Pathogens, Light Chemical Splash, Airborne Solid Particulates
Product Type Shoe Covers