Learn More
Ansell™ AlphaTec™ 3000 AVANT2 AIRline Ultrasonically Welded and Taped
Product Code. 16874990
Change view
Click to view available options
Ankle Style:
Attached Socks with Elasticated Boot Overflap
Unit Size:
Pack of 4
Product Code. Ankle Style Unit Size
16874990 Attached Socks with Elasticated Boot Overflap Pack of 4
1 options
Product Code. 16874990 Supplier Ansell™ Supplier No. YE30T00075602
Estimated Shipment: 27-05-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
AlphaTec™ 3000 AVANT2 AIRline Ultrasonically welded and taped is an encapsulated suit designed for use in combination with the permanently attached AlphaTec™ and AVANT STS V.1 airline regulator.
Non-gas-tight encapsulated suit designed to be used with the permanently attached AlphaTec™ and AVANT STS V.1 airline regulator.
  • Includes internal air-channel ventilation system 
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Welded and taped seams, our highest barrier to liquids and particulates
  • Panoramic visor design ensures a good field of vision for the wearer
  • A range of sleeve options available, including double cuffs and permanently attached chemical protective gloves
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow)
  • Suit/Belt mounted regulator complete with a low flow warning whistle the AVANT STS regulator can be mounted on the suit and removed for reuse providing cleaning and decontamination is permissible.
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
  • Attached socks with elasticated boot overflap - Socks are designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5)
Color Yellow
For Use With (Application) Opening and draining pumps valves or lines Filling, blending and charging of raw materials Contaminated Environments Chemical Handling Transferring liquids and solids Loading and unloading of proces equipment Raw material sample collection Concentration mixing and palletisation Hazmat response Assembly and disassembly of large pipes
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size Small
Ankle Style Attached Socks with Elasticated Boot Overflap
Closure Type Zipper
Cuff Style Double cuffs, finger and thumb loop
Disposable Reusable
Gender Unisex
Neck Style Encapsulated Hood
No. of Pockets None
Resistance CE Category III: The highest level of personal protective equipment (PPE) for severe risks. Type 3: Protective clothing that shields against liquid splashes. EN 1073-1: Standard for protective clothing against radioactive contamination. EN 1149-5: Standard for protective clothing with electrostatic properties, reducing static electricity discharges.
Seam Type ULTRASONICALLY WELDED AND TAPED
Sterility Non-sterile