missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 3000 Avant2 Airline Ultrasonically Welded and Taped - Model 757
Description
- Specialized protection: The AlphaTec™ 3000 AVANT2 AIRline Ultrasonically Welded and Taped - Model 757 full body protection suit has been tested against 200 chemicals.
- Genuine versatility: This chemical protective suit's HEPA filter offers secondary protection from airline contamination and a SMC silencer maintains the noise within a suit below 70 dB (even at maximum airflow)
- Considered aesthetics: In addition, this protective suit has attached socks with an elasticated boot overflap and is designed to be worn inside chemical protective boots with the leg overflap worn outside to reduce the potential for chemical ingress
- Reduced risks: The emergency ripcord feature of this full protective suit permits quick doffing of the suit in cases of emergency or undue distress to the wearer
- Fully encapsulated suit designed for use in combination with the permanently attached AlphaTec™ AVANT STS V.1 airline regulator.
- Suit features include internal air-channel ventilation system double cuffs, finger and thumb loop, G01 Ansell Solvex (Nitrile) gloves, G02 Ansell Barrier gloves or G05 AlphaTec™ 29-500 gloves.
- Attached socks with elasticated boot over-flap and emergency rip cord.
Spezifikation
Spezifikation
| Certifications/Compliance | CE CATEGORY III (Type 3, EN 1073-1, EN 1149-5) |
| Color | Yellow |
| For Use With (Application) | Opening and draining pumps valves or lines, Filling, blending and charging of raw materials, Contaminated Environments, Chemical Handling, Transferring liquids and solids, Raw material sample collection, Concentration mixing and palletisation, Hazmat response, Assembly and disassembly of large pipes, Loading and unloading of proces equipment |
| Hood Type | Fully Encapsulated |
| Material | Multi-layer Barrier Laminate Fabric |
| Size | Large |
| Ankle Style | Attached Socks, Elasticated Boot Over Flap |
| Disposable | Reusable |
| For Use With (Equipment) | Air-Fed/Supplied Systems |
| Gender | Unisex |
| Mehr anzeigen |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?