Learn More
Ansell AlphaTec™ 3000 AVANT2 AIRline
Product Code. p-7228387
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 4
Product Code. 17141065
Brand: Ansell YE30T00075703G05
Only null left
Add to basket
Add to basket
Encapsulated chemical protective suits for use with air-fed/supplied systems.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Panoramic visor design ensures a good field of vision for the wearer.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow). 
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer.
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
For Use With (Application) Transferring Liquids and Solids Between Vessels and Tanks and Process Equipment, Opening and Draining Pumps Valves or Lines, Filling, Blending and Charging of Raw Materials, Mixing or Loading Chemicals, Cleaning of Plant and Machines, Unexpected Leakages Spills or Other Releases, Loading and Unloading Trucks and Vehicles, Product Inspection
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size Medium
Product Type Encapsulated chemical protective suits for use with air-fed/supplied systems
Seam Type Welded and Taped
Style Y300757D
Weight (Metric) 80 gsm