Learn More
Ansell AlphaTec™ 3000 AVANT2 AIRline
Product Code. 17161065
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 4
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17161065 Large Pack of 4
17141065 Medium Pack of 4
17181065 X-Large Pack of 4
17121065 Small Pack of 4
17101075 2X-Large Pack of 4
17121075 3X-Large Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17161065 Supplier Ansell Supplier No. YE30T00075704G05
Add to basket
Add to basket
Encapsulated chemical protective suits for use with air-fed/supplied systems.
  • Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
  • Panoramic visor design ensures a good field of vision for the wearer.
  • HEPA filter provides secondary protection from airline contamination and a SMC silencer ensures the noise inside the suit is always below 70 dB (even at maximum airflow). 
  • Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer.
TRUSTED_SUSTAINABILITY
Specifications
Color Yellow
For Use With (Application) Transferring Liquids and Solids Between Vessels and Tanks and Process Equipment, Opening and Draining Pumps Valves or Lines, Filling, Blending and Charging of Raw Materials, Mixing or Loading Chemicals, Cleaning of Plant and Machines, Unexpected Leakages Spills or Other Releases, Loading and Unloading Trucks and Vehicles, Product Inspection
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate Fabric
Size Large
Product Type Encapsulated chemical protective suits for use with air-fed/supplied systems
Seam Type Welded and Taped
Style Y300757D
Weight (Metric) 80 gsm