Learn More
Ansell™ AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and Taped
Product Code. 31114778 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size
31114778 X-Large
31114775 Small
31114776 Medium
31114777 Large
31114779 2X-Large
31114780 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 31114778 Supplier Ansell™ Supplier No. YE30T0021405
Safety Starts Here: Save Up to 60%
Use promo code 28008
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Multi-layer chemical barrier, protects against numerous chemicals and enhances workplace safety
  • The AlphaTec™ 3000 Apron with Sleeves Ultrasonically Welded and taped - Model 214 chemical protective suit is made with a multi-layer barrier fabric, which offers wearers excellent protection against several chemicals, including inorganic and biological hazards
  • These highly visible, bright yellow chemical protection coveralls improve worker safety in dimly lit environments
  • Featuring welded and taped seams, this full chemical protection suit offers a strong barrier against harmful liquid and particles
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size X-Large
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Single-use
Closure Type Hook and Loop (Rear)
Sterility Non-sterile
Latex Free Yes
No. of Pockets None
Seam Type Welded and Taped
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, Type PB [6]-B, EN 14126 / ASTM F1671 - For US only)