Learn More
Ansell Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded - Model 243-G01
Product Code. 16854940
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Unit Size:
Pack of 25
Pack of 30
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16854940 X-Large Pack of 30
16824940 Small Pack of 30
16834940 Medium Pack of 30
16844940 Large Pack of 30
16864940 2X-Large Pack of 25
16874940 3X-Large Pack of 25
16884940 4X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 16854940 Supplier Ansell Supplier No. YE30T0024305G01
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded has rear velcro fastening at the neck.
DESIGN FEATURES:
  • Apron with sleeves with rear velcro fastening at the neck and attached AlphaTec™ 37-676
RECOMMENDED FOR:
  • Unexpected leakages spills or other releases
  • Loading and unloading trucks and vehicles
  • Loading trucks and vehicles
  • Product inspection
  • Cleaning of plant and machines
TRUSTED_SUSTAINABILITY
Specifications
Type Chemical-resistant Apron
Size X-Large
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Reusable
Closure Type Hook and Loop
Sterility Non-sterile
Weight (Metric) 80 gsm
Seam Type Welded
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126)