Learn More
Ansell™ AlphaTec™ 3000 Apron - Model 213
Product Code. 31114774 Shop All Ansell Products
Change view
Click to view available options
Closure Type:
Ties
Product Code. Closure Type
31114774 Ties
1 options
Product Code. 31114774 Supplier Ansell™ Supplier No. YE30T0021300
Estimated Shipment: 13-05-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protective multi-layer barrier, protection against numerous chemicals with Lightweight and durable material
  • This AlphaTec™ chemical-protective apron features a multi-layer fabric that protects against various inorganic chemicals and biological hazards
  • Bright yellow color, ensures highly noticeable personal protection, enhancing workplace safety and safeguarding worker welfare
  • An extremely light protective apron, the AlphaTec™ 3000 Apron - Model 213 remains highly comfortable, even with extended wear
  • Its multi-layer fabric construction also means this chemical-resistant apron withstands wear and tear
TRUSTED_SUSTAINABILITY
Specifications
Type Apron
Size Universal
Material Multi-layer Barrier Laminate Fabric
Color Yellow
Disposable Single-use
Closure Type Ties
Sterility Non-sterile
Latex Free Yes
No. of Pockets None
Seam Type Seamless
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, Type PB [6]-B, EN 14126)