Learn More
Ansell AlphaTec™ 2000 STANDARD Oversleeves Bound
Product Code. 16844800
Click to view available options
Size:
Universal
X-Large
Unit Size:
250 pairs
300 pairs
Product Code. 16844800
Brand: Ansell WH20B0060005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Comfortable oversleeves featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Material Microporous Non-Woven Polyethylene Laminate
Color White
Seam Type Bound
Model WH20B
For Use With (Application) Cleanroom Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services
Certifications/Compliance (Europe) EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013, Partial Body Protection Only
Weight (Metric) 63 gsm