missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD Jacket & Trouser Set Bound
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Optimized fit, improving wearer comfort and safety
- Low Linting
Specifications
Specifications
| Size | 2X-Large |
| Gender | Unisex |
| Material | Microporous Non-woven Polyethylene Laminate |
| Color | White |
| Closure Type | Zipper |
| Seam Type | Bound |
| Certifications/Compliance (Europe) | Partial Body Protection Only, EN 13034:2005 + A1:2009, DIN 32781:2010, EN 14126:2003, ASTM F1671, EN ISO 13688:2013 |
| Product Type | Chemical-resistant Jacket |
| Style | W20B219 |
| For Use With (Application) | Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?