Learn More
Ansell Alphatec™ 1500 Plus Lab Coat Bound - Model 206
Product Code. 17190995
Change view
Click to view available options
Size:
X-Large
2X-Large
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17190995 X-Large Pack of 50
17101005 2X-Large Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17190995 Supplier Ansell Supplier No. WH15B0020605
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
Alphatec™ 1500 Plus Lab Coat delivers comfortable protection in general industrial environments.
  • Optimized breathability: AlphaTec™ 1500 PLUS Lab Coat Bound - Model 204 delivers comfortable protection in general industrial environments, thanks to its air and water vapor permeable as well as anti-static, SMS material
  • Reduced contamination risks: As this protective outer garment is silicone-free, it minimizes contamination risks, making it ideal for the life sciences and metal fabrication industries
  • Improved ergonomics: In addition, this white protective over garment offers an optimized fit, which improves wearer comfort and workplace safety
DESIGN FEATURES:
  • Medium length labcoat with collar, studded front fastening, left breast and right thigh pockets.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Anti-static Lab Coat
Size X-Large
Gender Unisex
Material SMS
Color White
Closure Type Buttons
Cuff Style Elastic
Disposable Reusable
Sterility Non-sterile
Resistance Chemicals
Style Full-length
No. of Pockets 2
Latex Free Yes
Certifications/Compliance (Europe) CE CATEGORY III (Type PB [6])
Weight (English) 48 gsm
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.