missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 04-005 Certified Heavy Duty PVC Chemical Protection Gloves - Extended Cuff

Product Code. 16659063 Shop All Ansell Products
Change view
Click to view available options
Size:
9
10
11
Unit Size:
12 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
16659063 10 12 pairs
16649063 9 12 pairs
16669063 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16659063 Supplier Ansell Supplier No. 04005100
Only null left
Add to basket
Edge
Add to basket

A specialty blended formulation of nitrile and PVC provides durability, flexibility, and protection. Gloves are EN ISO 374:2016 Type A Certified to protect against a variety of chemicals.

  • Nitrile and PVC blend provides chemical protection against 6 chemicals certified (JKLMPT) out of 18 EN ISO 374:2016
  • An additional 5 chemicals (AFNOS) have achieved a permeation level 2 or higher in an accredited lab
  • Special foam design provides oil resistance, and enhances grip capabilities in wet or oily environments
  • 13-gauge seamless cotton design increases comfort and protection
  • Complies with FDA and EU food regulations
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009
Applications Industrial
Color Blue
Latex Free No
Material Nitrile/PVC Blend
Product Type Chemical-resistant Gloves
Size 10
EN388 Standard 4131A
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES JKLMPT
EN ISO 374 - 5 Yes
Glove Cuff Style Gauntlet
Liner Material Cotton
Resistant To Oil and Various Chemicals
Series 04-005
Length (English) 15 in.
Length (Metric) 380 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.