missing translation for 'onlineSavingsMsg'
Learn More

(AlphaS)-Alpha-[[(1S)-1-Carboxyethyl]amino]benzenebutanoic Acid, TRC

Product Code. 30354821
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354821 100 mg 100mg
30350182 25 mg 25mg
30350312 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354821 Supplier Toronto Research Chemicals Supplier No. TRCC178243100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84023-58-5
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.28
Synonym [S-(R*,R*)]-alpha-[(1-Carboxyethyl)amino]-benzenebutanoic Acid
IUPAC Name (2S)-2-[[(1S)-1-carboxyethyl]amino]-4-phenylbutanoic acid
SMILES C[C@H](N[C@@H](CCc1ccccc1)C(=O)O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H17NO4/c1-9(12(15)16)14-11(13(17)18)8-7-10-5-3-2-4-6-10/h2-6,9,11,14H,7-8H2,1H3,(H,15,16)(H,17,18)/t9-,11-/m0/s1
Formula Weight 251.1158
Chemical Name or Material (alphaS)-alpha-(((1S)-1-Carboxyethyl)amino)benzenebutanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.