missing translation for 'onlineSavingsMsg'
Learn More

(AlphaS)-Alpha-[[(1,1-Dimethylethoxy)carbonyl]amino]benzenebutanoyl-L-leucyl-phenylmethyl Ester L-Phenylalanine, TRC

Product Code. 30385267
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385267 250 mg
30381249 500 mg
30386612 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385267 Supplier Toronto Research Chemicals Supplier No. TRCD465510250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 868540-15-2
Molecular Formula C37H47N3O6
Molecular Weight (g/mol) 629.79
Synonym (As)-A-[[(1,1-Dimethylethoxy)Carbonyl]Amino]Benzenebutanoyl-L-Leucyl-Phenylmethyl Ester L-Phenylalanine
IUPAC Name benzyl (2R)-2-[[(2S)-4-methyl-2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoyl]amino]pentanoyl]amino]-3-phenylpropanoate
SMILES CC(C)C[C@H](NC(=O)C(CCc1ccccc1)NC(=O)OC(C)(C)C)C(=O)N[C@H](Cc2ccccc2)C(=O)OCc3ccccc3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C37H47N3O6/c1-26(2)23-31(38-33(41)30(40-36(44)46-37(3,4)5)22-21-27-15-9-6-10-16-27)34(42)39-32(24-28-17-11-7-12-18-28)35(43)45-25-29-19-13-8-14-20-29/h6-20,26,30-32H,21-25H2,1-5H3,(H,38,41)(H,39,42)(H,40,44)/t30?,31-,32+/m0/s1
Formula Weight 629.3465
Chemical Name or Material (alphaS)-alpha-(((1,1-Dimethylethoxy)carbonyl)amino)benzenebutanoyl-L-leucyl-phenylmethyl Ester L-Phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.