missing translation for 'onlineSavingsMsg'
Learn More

(AlphaR)-Alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxy-benzeneacetic Acid, TRC

Product Code. 30385046
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385046 25 g
30384608 50 g
30386180 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385046 Supplier Toronto Research Chemicals Supplier No. TRCE91736525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62893-24-7
Molecular Formula C15H17N3O6
Molecular Weight (g/mol) 335.31
Synonym (R)-alpha-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]-4-hydroxy-benzeneacetic Acid, HO-EPCP
IUPAC Name (R)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)-2-(4-hydroxyphenyl)acetic acid
SMILES O=C(N(CCN(CC)C1=O)C1=O)N[C@H](C2=CC=C(O)C=C2)C(O)=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C15H17N3O6/c1-2-17-7-8-18(13(21)12(17)20)15(24)16-11(14(22)23)9-3-5-10(19)6-4-9/h3-6,11,19H,2,7-8H2,1H3,(H,16,24)(H,22,23)/t11-/m1/s1
Formula Weight 335.11
Chemical Name or Material (alphaR)-alpha-(((4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl)amino)-4-hydroxy-benzeneacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.