missing translation for 'onlineSavingsMsg'
Learn More

Alphacel Non-Nutritive Bulk, MP Biomedicals

Product Code. 11451392 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 lb.
100 lb.
Unit Size:
100lb
5lb
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11451392 100 lb. 100lb
11431392 5 lb. 5lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451392 Supplier MP Biomedicals Supplier No. 0290045380
  • ONLINE EXCLUSIVE PRICE
    £989.00
    £927.15 / 100lb
    Save £61.85
    6% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Composed of finely ground cellulose for addition to animal diets to supply bulk with minimum nutritional value

Chemical Identifiers

CAS 9004-34-6

Specifications

Quantity 100 lb.
Chemical Name or Material Alphacel Non-Nutritive Bulk
compliance-icons
Product Identifier
  • Alphacel
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Alphacel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.