missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaB Crystallin/CRYAB, p Ser45 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-5598-0.025mg
This item is not returnable.
View return policy
Description
AlphaB Crystallin/CRYAB Polyclonal specifically detects AlphaB Crystallin/CRYAB in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| AlphaB Crystallin/CRYAB (p Ser45) | |
| Polyclonal | |
| Western Blot 0.5 μg/mL, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 8 μg/mL, Immunofluorescence 5 μg/mL | |
| alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This does not detect the unphosphorylated form of the protein. | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CRYAB | |
| Synthetic phopshopeptide corresponding to residues L(44) S(p) P F Y L R P P S F(54) C of human alpha-B Crystallin. | |
| 0.025 mg | |
| Neuroscience, Phospho Specific, Vision | |
| 1410 | |
| Reconstitute to desired concentration. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction