missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaB Crystallin/CRYAB Antibody (CRYAB/7918), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20421-100ug
This item is not returnable.
View return policy
Description
AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| AlphaB Crystallin/CRYAB | |
| Monoclonal | |
| 0.2 mg/ml | |
| Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component | |
| Recombinant human full-length AlphaB Crystallin/CRYAB protein | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision | |
| 1410 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CRYAB/7918 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction