missing translation for 'onlineSavingsMsg'
Learn More

AlphaB Crystallin/CRYAB Antibody (CRYAB/4663), Novus Biologicals™

Mouse Monoclonal Antibody

£195.00 - £385.00

Specifications

Antigen AlphaB Crystallin/CRYAB
Clone CRYAB/4663
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18612504
View Documents
Novus Biologicals
NBP3-20407-100ug
100 μg
£385.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18641594
View Documents
Novus Biologicals
NBP3-20407-20ug
20 μg
£195.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

AlphaB Crystallin/CRYAB
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision
10mM PBS with 0.05% BSA
1410
IgG1 κ
Protein A or G purified
CRYAB/4663
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
Human, Rat
alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
Recombinant human full-length AlphaB Crystallin/CRYAB protein
Primary
Store at 4&drg;C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.