Learn More
AlphaB Crystallin/CRYAB Antibody (CL10758) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21180AF488
Product Code. 30296397
  • £558.00 / 0.10mL
Description
AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AlphaB Crystallin/CRYAB
Monoclonal
Alexa Fluor 488
alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
This antibody was developed using a synthetic peptide derived from P02511, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Phospho Specific, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision
1410
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL10758
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Rat
Purified