missing translation for 'onlineSavingsMsg'
Learn More

AlphaB Crystallin/CRYAB Antibody (3A10.C9), DyLight 650, Novus Biologicals™

Product Code. 18432543 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432543 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432543 Supplier Novus Biologicals Supplier No. NBP122410C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AlphaB Crystallin/CRYAB Monoclonal antibody specifically detects AlphaB Crystallin/CRYAB in Human, Rat, Bovine samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen AlphaB Crystallin/CRYAB
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 3A10.C9
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Accession No. P02511, NP_001876.1
Gene Alias alpha B crystallin, alpha(B)-crystallin, alpha-crystallin B chain, CRYA2alpha crystallin B chain, crystallin, alpha B, CTPP2, Heat shock protein beta-5, heat-shock 20 kD like-protein, HSPB5, Renal carcinoma antigen NY-REN-27, Rosenthal fiber component
Host Species Mouse
Immunogen Native Alpha B Crystallin
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience, Neurotransmission, Protein Phosphatase, Sensory Systems, Signal Transduction, Tyrosine Kinases, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1410
Target Species Human, Rat, Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.