missing translation for 'onlineSavingsMsg'
Learn More

AlphaA Crystallin/CRYAA Antibody (OTI3B12), DyLight 594, Novus Biologicals™

Product Code. 18422413 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18422413 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18422413 Supplier Novus Biologicals Supplier No. NBP270450DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AlphaA Crystallin/CRYAA Monoclonal antibody specifically detects AlphaA Crystallin/CRYAA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen AlphaA Crystallin/CRYAA
Applications Western Blot, Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone OTI3B12
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4
Host Species Mouse
Immunogen Full length recombinant protein of human AlphaA Crystallin/CRYAA (NP_000385) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Membrane Trafficking and Chaperones, Vision
Primary or Secondary Primary
Gene ID (Entrez) 1409
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.