missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaA Crystallin/CRYAA Antibody (c9F2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-14821
This item is not returnable.
View return policy
Description
AlphaA Crystallin/CRYAA Monoclonal specifically detects AlphaA Crystallin/CRYAA in Human, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| AlphaA Crystallin/CRYAA | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:2000, ELISA 1:100-1:2000 | |
| CRYA1crystallin, alpha-1, crystallin, alpha A, Heat shock protein beta-4, HSPB4alpha-crystallin A chain, human alphaA-crystallin (CRYA1), 10HspB4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB120-14821 recognizes alpha A crystallin, a 20kDa water-soluble major structural protein and member of the heat shock protein family, designated HSPB4, which acts as a molecular chaperone and is believed to play a role in the maintenance of ocular lens transparency/refractive index. Besides the involvement of both alpha A and alpha B crystallin in the prevention of protein precipitation and cataract formation in the eye, alpha crystallins have also been demonstrated in other tissues including the heart, brain, kidney, spleen and thymus. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| c9F2 | |
| Unconjugated | |
| P02489 | |
| CRYAA | |
| E. coli-derived recombinant protein corresponding to full-length (aa1-173) human crystallin alpha A. | |
| 0.1 mL | |
| Membrane Trafficking and Chaperones, Vision | |
| 1409 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction