missing translation for 'onlineSavingsMsg'
Learn More

(Alpha1S,Alpha4S,2S,5S)-2,5-Dimethyl-3,6-dioxo-Alpha1,Alpha4-bis(2-phenylethyl)-1,4-piperazinediacetic Acid 1,4-Diethyl Ester, TRC

Product Code. 30375672
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375672 1 g
30385561 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30375672 Supplier Toronto Research Chemicals Supplier No. TRCD4710501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151387-05-2
Molecular Formula C30H38N2O6
Molecular Weight (g/mol) 522.63
Synonym [2S-[1(R*),2alpha,4(R*),5alpha]]-2,5-Dimethyl-3,6-dioxo-alpha,alpha'-bis(2-phenylethyl)-1,4-piperazinediacetic Acid Diethyl Ester, Ramipril Impurity O
SMILES CCOC(=O)[C@H](CCc1ccccc1)N2[C@@H](C)C(=O)N([C@@H](CCc3ccccc3)C(=O)OCC)[C@@H](C)C2=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C30H38N2O6/c1-5-37-29(35)25(19-17-23-13-9-7-10-14-23)31-21(3)28(34)32(22(4)27(31)33)26(30(36)38-6-2)20-18-24-15-11-8-12-16-24/h7-16,21-22,25-26H,5-6,17-20H2,1-4H3/t21-,22-,25-,26-/m0/s1
Formula Weight 522.273
Chemical Name or Material (alpha1S,alpha4S,2S,5S)-2,5-Dimethyl-3,6-dioxo-alpha1,alpha4-bis(2-phenylethyl)-1,4-piperazinediacetic Acid 1,4-Diethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.